4-(Hydroxymethyl)benzoesäure
Lieferant: NOVABIOCHEM
Synonyme:
4-Carboxybenzylalkohol, HMBA
8.51042.0025
8.51042.0005
01-60-0007-25EA
566
EUR
01-60-0007-25
01-60-0007-5
4-(Hydroxymethyl)benzoesäure
4-(Hydroxymethyl)benzoic acid
Formel:
C₈H₈O₃ Molecular Weight: 152,15 g/mol |
MDL:
MFCD00017598 CAS-Nummer: 3006-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...