4-(Hydroxymethyl)benzoesäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Carboxybenzylalkohol, HMBA

M-Clarity™ Qualitätsstufe = MQ100

382639-10G 382639-1G
SIAL382639-10GEA 131 EUR
SIAL382639-10G SIAL382639-1G
4-(Hydroxymethyl)benzoesäure, Sigma-Aldrich®
4-(Hydroxymethyl)benzoic acid
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
MDL: MFCD00017598
CAS-Nummer: 3006-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR