4-(Trifluormethoxy)benzolsulfonamid

Lieferant: FLUOROCHEM

Synonyme: 4-(Trifluormethoxy)benzolsulfonsäureamid

CFF1279. 005531-1G 005531-5G
CHMPCFF1279.EA 0 EUR
CHMPCFF1279. FLUO005531-1G FLUO005531-5G
4-(Trifluormethoxy)benzolsulfonamid
4-(Trifluormethoxy)benzolsulfonamid
Formel: C₇H₆F₃NO₃S
Molecular Weight: 241.19 g/mol
MDL: MFCD00798381
CAS-Nummer: 1513-45-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR