4,4,5,5,5-Pentafluor-1-pentanol

Lieferant: FLUOROCHEM

Synonyme: 4,4,5,5,5-Pentafluoropentanol

006963-5G 006963-25G 006963-100G
FLUO006963-5GEA 73.1 EUR
FLUO006963-5G FLUO006963-25G FLUO006963-100G
4,4,5,5,5-Pentafluor-1-pentanol
4,4,5,5,5-Pentafluoropentan-1-ol
Formel: C₅H₇F₅O
Molecular Weight: 178,1 g/mol
MDL: MFCD00153224
CAS-Nummer: 148043-73-6
EINECS: 421-360-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR