4,4-Dimethylcyclohexanon
Lieferant: COMBI-BLOCKS
Synonyme:
4,4-Dimethylcyclohexan-1-one
CS-9107-5G
CS-9107-1G
COBBCS-9107-5GEA
115
EUR
COBBCS-9107-5G
COBBCS-9107-1G
4,4-Dimethylcyclohexanon
4,4-Dimethylcyclohexanone
Formel:
C₈H₁₄O Molecular Weight: 126,2 g/mol |
MDL:
MFCD00234965 CAS-Nummer: 4255-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...