4,5-Dicyanimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
Imidazol-4,5-dicarbonitril, 4,5-Dicyan-1H-imidazol
HI-1161-5G
HI-1161-25G
HI-1161-100G
COBBHI-1161-5GEA
0
EUR
COBBHI-1161-5G
COBBHI-1161-25G
COBBHI-1161-100G
4,5-Dicyanimidazol
4,5-Dicyanoimidazole
Formel:
C₅H₂N₄ Schmelzpunkt: 176 °C Dichte: 1,458 g/cm³ (20 °C) |
MDL:
MFCD00005194 CAS-Nummer: 1122-28-7 EINECS: 214-344-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...