4,5-Dicyanimidazol
Lieferant: CHEMPUR
Synonyme:
Imidazol-4,5-dicarbonitril, 4,5-Dicyan-1H-imidazol
FL13519.POR
013519-25G
CHMPFL13519.POREA
0
EUR
CHMPFL13519.POR
FLUO013519-25G
4,5-Dicyanimidazol
4,5-Dicyanoimidazole
Formel:
C₅H₂N₄ Schmelzpunkt: 176 °C Dichte: 1,458 g/cm³ (20 °C) |
MDL:
MFCD00005194 CAS-Nummer: 1122-28-7 EINECS: 214-344-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...