4,5-Diiod-2-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Diiod-2-methyl-1H-imidazol

HI-1059-1G HI-1059-25G HI-1059-5G
COBBHI-1059-1GEA 93.7 EUR
COBBHI-1059-1G COBBHI-1059-25G COBBHI-1059-5G
4,5-Diiod-2-methylimidazol
4,5-Diiodo-2-methylimidazole
Formel: C₄H₄I₂N₂
Molecular Weight: 333,9 g/mol
MDL: MFCD00464337
CAS-Nummer: 73746-44-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR