4,5-Diiodimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Diiod-1H-imidazol

HI-1002-5G HI-1002-25G HI-1002-1G
COBBHI-1002-5GEA 63.4 EUR
COBBHI-1002-5G COBBHI-1002-25G COBBHI-1002-1G
4,5-Diiodimidazol
4,5-Diiodoimidazole
Formel: C₃H₂I₂N₂
Molecular Weight: 319,87 g/mol
MDL: MFCD00963878
CAS-Nummer: 15813-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR