Chrysin, gelbes Pulver
Lieferant: MP Biomedicals
Synonyme:
5,7-Dihydroxy-2-phenyl-4H-chromen-4-on, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-on, 5,7-Dihydroxy-2-phenyl-4-benzopyron, 5,7-Dihydroxy-2-phenylchromen-4-on, 5,7-Dihydroxyflavon
0215498525
0215498505
ICNA0215498525EA
150
EUR
ICNA0215498525
ICNA0215498505
Chrysin, gelbes Pulver
Chrysin
Soluble in pyridine (50 mg/ml) (Lit.)
Formel:
C₁₅H₁₀O₄ Molecular Weight: 254,24 g/mol Siedepunkt: 429 °C (1013 hPa) Schmelzpunkt: 284…290 °C (Lit.) |
MDL:
MFCD00006834 CAS-Nummer: 480-40-0 EINECS: 207-549-7 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...