Chrysin ≥98%
Lieferant: Thermo Scientific
Synonyme:
5,7-Dihydroxy-2-phenyl-4H-chromen-4-on, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-on, 5,7-Dihydroxy-2-phenyl-4-benzopyron, 5,7-Dihydroxy-2-phenylchromen-4-on, 5,7-Dihydroxyflavon
L14178.06
L14178.14
L14178.06EA
32.1
EUR
L14178.06
L14178.14
Chrysin ≥98%
Chrysin
Purity: 98%
CAS No.: 480-40-0
Molecular Formula: C15H10O4
Formula Weight: 254.24
Melting Point: 284-289°
Beilstein Reference: 233276
Merck Reference: 14,2256
MDL No.: MFCD00006834
Formel:
C₁₅H₁₀O₄ Molecular Weight: 254,24 g/mol Siedepunkt: 429 °C (1013 hPa) Schmelzpunkt: 284…289 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006834 CAS-Nummer: 480-40-0 EINECS: 207-549-7 Merck Index: 14,2256 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...