Chrysin 99+%
Lieferant: Thermo Scientific
Synonyme:
5,7-Dihydroxy-2-phenyl-4H-chromen-4-on, 5,7-Dihydroxy-2-phenyl-4H-benzo[b]pyran-4-on, 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-on, 5,7-Dihydroxy-2-phenyl-4-benzopyron, 5,7-Dihydroxy-2-phenylchromen-4-on, 5,7-Dihydroxyflavon
110320050
110320250
110321000
ACRO110320050EA
53
EUR
ACRO110320050
ACRO110320250
ACRO110321000
Chrysin 99+%
Chrysin
Formel:
C₁₅H₁₀O₄ Molecular Weight: 254,24 g/mol Siedepunkt: 429 °C (1013 hPa) Schmelzpunkt: 284…290 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006834 CAS-Nummer: 480-40-0 EINECS: 207-549-7 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 284 to 290 °C |
Loss on drying | ≤1.0 % |
HPLC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...