Genistein
Lieferant: Cayman Chemical
Synonyme:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-on, 4',5,7-Trihydroxyisoflavon, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyron
10005167-100
10005167-250
10005167-500
10005167-1
CAYM10005167-100EA
42
EUR
CAYM10005167-100
CAYM10005167-250
CAYM10005167-500
CAYM10005167-1
Genistein
Genistein
Formel:
C₁₅H₁₀O₅ Molecular Weight: 270,24 g/mol Schmelzpunkt: 297…298 °C |
MDL:
MFCD00016952 CAS-Nummer: 446-72-0 EINECS: 207-174-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...