Biochanin A, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-1-benzopyran-4-on, 5,7-Dihydroxy-4'-methoxyisoflavon, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4H-chromen-4-on, 5,7-Dihydroxy-3-(4-methoxyphenyl)chromen-4-on, 5,7-Dihydroxy-3-(4-methoxyphenyl)-4-benzopyron, 4'-Methoxy-5,7-dihydroxyisoflavon

M-Clarity™ Qualitätsstufe = MQ100

D2016-100MG D2016-1G D2016-250MG
SIALD2016-100MGEA 83.3 EUR
SIALD2016-100MG SIALD2016-1G SIALD2016-250MG
Biochanin A, Sigma-Aldrich®
Biochanin A
Formel: C₁₆H₁₂O₅
Molecular Weight: 284,27 g/mol
MDL: MFCD00006839
CAS-Nummer: 491-80-5
EINECS: 207-744-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR