4,8-Dichlorchinolin

Lieferant: COMBI-BLOCKS
Achtung

HC-2310-5G HC-2310-1G
COBBHC-2310-5GEA 282 EUR
COBBHC-2310-5G COBBHC-2310-1G
4,8-Dichlorchinolin
4,8-Dichloroquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198,05 g/mol
MDL: MFCD00024008
CAS-Nummer: 21617-12-9
EINECS: 244-480-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR