4-Amino-3-brom-2-chlorpyridin ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Brom-2-chlor-4-pyridylamin

H52359.03 H52359.06
H52359.03EA 135 EUR
H52359.03 H52359.06
4-Amino-3-brom-2-chlorpyridin ≥97%
4-Amino-3-bromo-2-chloropyridine

Purity: 97+% CAS No.: 215364-85-5 Molecular Formula: C5H4BrClN2 Formula Weight: 207.46 MDL No.: MFCD09263478

Formel: C₅H₄BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09263478
CAS-Nummer: 215364-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR