4-Amino-3-brompyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Brompyridin-4-amin, 3-Brom-4-pyridylamin

OS-7639-5G OS-7639-1G OS-7639-25G
COBBOS-7639-5GEA 38.2 EUR
COBBOS-7639-5G COBBOS-7639-1G COBBOS-7639-25G
4-Amino-3-brompyridin
4-Amino-3-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173,01 g/mol
Siedepunkt: 244 °C (1013 hPa)
Schmelzpunkt: 181…182,5 °C
MDL: MFCD02068297
CAS-Nummer: 13534-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR