4-Amino-3-fluorbenzoesäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
4-Amino-3-fluorobenzoic Acid
CA-4027-5G
CA-4027-25G
CA-4027-1G
COBBCA-4027-5GEA
35.9
EUR
COBBCA-4027-5G
COBBCA-4027-25G
COBBCA-4027-1G
4-Amino-3-fluorbenzoesäure
4-Amino-3-fluorobenzoic acid
Formel:
C₇H₆FNO₂ Molecular Weight: 155.13 g/mol |
MDL:
MFCD01660374 CAS-Nummer: 455-87-8 EINECS: 207-250-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...