4-Amino-3-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 3-Methylpyridin-4-amin, 4-Amino-3-picolin, 3-Methyl-4-pyridylamin

PY-7853-1G PY-7853-25G PY-7853-5G
COBBPY-7853-1GEA 26.2 EUR
COBBPY-7853-1G COBBPY-7853-25G COBBPY-7853-5G
4-Amino-3-methylpyridin
4-Amino-3-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108,14 g/mol
MDL: MFCD01704431
CAS-Nummer: 1990-90-5
EINECS: 217-872-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR