4-Aminonicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure

CA-4067-5G CA-4067-25G CA-4067-1G
COBBCA-4067-5GEA 50.9 EUR
COBBCA-4067-5G COBBCA-4067-25G COBBCA-4067-1G
4-Aminonicotinsäure
4-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00234183
CAS-Nummer: 7418-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR