4-Aminobenzamidin Dihydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L14089.03
L14089.06
L14089.03EA
32.7
EUR
L14089.03
L14089.06
4-Aminobenzamidin Dihydrochlorid 97%
4-Aminobenzamidin Dihydrochlorid
Purity: 97%
CAS No.: 2498-50-2
Molecular Formula: C7H9N3.2HCl
Formula Weight: 208.09
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
C₇H₁₁Cl₂N₃ Schmelzpunkt: >300 °C |
MDL:
MFCD00013001 CAS-Nummer: 2498-50-2 EINECS: 219-692-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...