4-Amino-N,N-dimethylbenzolsulfonamid

Lieferant: BIONET RESEARCH

Synonyme: Anilin-4-sulfonsäuredimethylamid, 4-Aminobenzolsulfonsäuredimethylamid

1C-033.500 1C-033.1 1C-033.5 1C-033.10 KO1C-033.0001
BIOT1C-033.500EA 0 EUR
BIOT1C-033.500 BIOT1C-033.1 BIOT1C-033.5 BIOT1C-033.10 ABCRKO1C-033.0001
4-Amino-N,N-dimethylbenzolsulfonamid
4-Amino-N,N-dimethylbenzolsulfonamid
Formel: C₈H₁₂N₂O₂S
Molecular Weight: 200.26 g/mol
MDL: MFCD00031428
CAS-Nummer: 1709-59-7
EINECS: 216-970-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR