4-Aminothiobenzamid
Lieferant: FLUOROCHEM
Synonyme:
4-Aminothiobenzoesäureamid
021309-1G
021309-2.5G
FLUO021309-1GEA
55.2
EUR
FLUO021309-1G
FLUO021309-2.5G
4-Aminothiobenzamid
4-Aminothiobenzamid
Formel:
C₇H₈N₂S Molecular Weight: 152,22 g/mol |
MDL:
MFCD00040927 CAS-Nummer: 4714-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...