4-Brom-1,2-dimethylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-1,2-dimethyl-1H-imidazol

HI-1175-5G HI-1175-1G
COBBHI-1175-5GEA 0 EUR
COBBHI-1175-5G COBBHI-1175-1G
4-Brom-1,2-dimethylimidazol
4-Bromo-1,2-dimethylimidazole
Formel: C₅H₇BrN₂
MDL: MFCD06659901
CAS-Nummer: 850429-59-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR