4-Brom-1-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-1-methyl-1H-imidazol

HI-1026-5G HI-1026-1G
COBBHI-1026-5GEA 445 EUR
COBBHI-1026-5G COBBHI-1026-1G
4-Brom-1-methylimidazol
4-Bromo-1-methylimidazole
Formel: C₄H₅BrN₂
Molecular Weight: 161 g/mol
MDL: MFCD01320501
CAS-Nummer: 25676-75-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR