4-Brom-2,6-difluorbenzoesäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
OS-7660-5G
OS-7660-25G
OS-7660-100G
COBBOS-7660-5GEA
38.8
EUR
COBBOS-7660-5G
COBBOS-7660-25G
COBBOS-7660-100G
4-Brom-2,6-difluorbenzoesäure
4-Bromo-2,6-difluorobenzoic acid
Formel:
C₇H₃BrF₂O₂ Molecular Weight: 237 g/mol |
MDL:
MFCD03094085 CAS-Nummer: 183065-68-1 EINECS: 457-940-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...