4-Brom-2-chlorbenzoesäure

Lieferant: COMBI-BLOCKS

CA-4187-5G CA-4187-25G
COBBCA-4187-5GEA 31.2 EUR
COBBCA-4187-5G COBBCA-4187-25G
4-Brom-2-chlorbenzoesäure
4-Bromo-2-chlorobenzoic acid
Formel: C₇H₄BrClO₂
MDL: MFCD00040903
CAS-Nummer: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR