4-Brom-2-chlorpyridin

Lieferant: COMBI-BLOCKS

CA-4322-1G CA-4322-25G CA-4322-5G
COBBCA-4322-1GEA 0 EUR
COBBCA-4322-1G COBBCA-4322-25G COBBCA-4322-5G
4-Brom-2-chlorpyridin
4-Bromo-2-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD03840756
CAS-Nummer: 73583-37-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR