4-Brom-2-methylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-2-methyl-1H-imidazol
HI-1111-5G
HI-1111-25G
HI-1111-100G
COBBHI-1111-5GEA
78.6
EUR
COBBHI-1111-5G
COBBHI-1111-25G
COBBHI-1111-100G
4-Brom-2-methylimidazol
4-Bromo-2-methylimidazole
Formel:
C₄H₅BrN₂ Molecular Weight: 161 g/mol |
MDL:
MFCD03411959 CAS-Nummer: 16265-11-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...