4-Brom-2-methylpyridin

Lieferant: COMBI-BLOCKS

OS-7471-25G OS-7471-100G OS-7471-5G
COBBOS-7471-25GEA 140 EUR
COBBOS-7471-25G COBBOS-7471-100G COBBOS-7471-5G
4-Brom-2-methylpyridin
4-Brom-2-methylpyridin
Formel: C₆H₆BrN
Molecular Weight: 172.02 g/mol
MDL: MFCD03788196
CAS-Nummer: 22282-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR