4-Brom-2-methylthiophen
Lieferant: COMBI-BLOCKS
OS-7668-25G
OS-7668-100G
OS-7668-1G
OS-7668-5G
COBBOS-7668-25GEA
193
EUR
COBBOS-7668-25G
COBBOS-7668-100G
COBBOS-7668-1G
COBBOS-7668-5G
4-Brom-2-methylthiophen
4-Bromo-2-methylthiophene
Formel:
C₅H₅BrS Molecular Weight: 177,06 g/mol |
MDL:
MFCD04116367 CAS-Nummer: 29421-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...