4-Brom-N-tert-butyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-3-(trifluormethyl)benzolsulfonsäure-tert-butylamid

HC-6475-5G HC-6475-1G
COBBHC-6475-5GEA 0 EUR
COBBHC-6475-5G COBBHC-6475-1G
4-Brom-N-tert-butyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-tert-butyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₁₁H₁₃BrF₃NO₂S
Molecular Weight: 360.19
MDL: MFCD09972162
CAS-Nummer: 1020253-06-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR