4-Brom-N-butyl-3-(trifluormethyl)benzolsulfonamid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-3-(trifluormethyl)benzolsulfonsäurebutylamid
HC-6473-5G
HC-6473-1G
COBBHC-6473-5GEA
451
EUR
COBBHC-6473-5G
COBBHC-6473-1G
4-Brom-N-butyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-butyl-3-(trifluormethyl)benzolsulfonamid
Formel:
C₁₁H₁₃BrF₃NO₂S Molecular Weight: 360,19 g/mol |
MDL:
MFCD09972161 CAS-Nummer: 1020253-05-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...