4-Brom-N-cyclohexyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6469-5G HC-6469-1G
COBBHC-6469-5GEA 0 EUR
COBBHC-6469-5G COBBHC-6469-1G
4-Brom-N-cyclohexyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N-cyclohexyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₁₃H₁₅BrF₃NO₂S
MDL: MFCD09972158
CAS-Nummer: 1020253-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR