4-Brom-N,N-dimethyl-3-(trifluormethyl)benzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-3-(trifluormethyl)benzolsulfonsäuredimethylamid

HC-6472-5G HC-6472-1G
COBBHC-6472-5GEA 0 EUR
COBBHC-6472-5G COBBHC-6472-1G
4-Brom-N,N-dimethyl-3-(trifluormethyl)benzolsulfonamid
4-Brom-N,N-dimethyl-3-(trifluormethyl)benzolsulfonamid
Formel: C₉H₉BrF₃NO₂S
MDL: MFCD10699660
CAS-Nummer: 939989-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR