4-Brom-3-(trifluormethyl)-1H-pyrazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-3-(trifluormethyl)-1H-pyrazol

HC-2046-5G HC-2046-25G HC-2046-1G
COBBHC-2046-5GEA 0 EUR
COBBHC-2046-5G COBBHC-2046-25G COBBHC-2046-1G
4-Brom-3-(trifluormethyl)-1H-pyrazol
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Formel: C₄H₂BrF₃N₂
Molecular Weight: 214.97 g/mol
MDL: MFCD04969230
CAS-Nummer: 19968-17-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR