4-Brom-3-fluorbenzolsulfonamid

Lieferant: FLUOROCHEM

013778-5G 013778-25G
FLUO013778-5GEA 63.9 EUR
FLUO013778-5G FLUO013778-25G
4-Brom-3-fluorbenzolsulfonamid
4-Brom-3-fluorbenzolsulfonamid
Formel: C₆H₅BrFNO₂S
MDL: MFCD03094292
CAS-Nummer: 263349-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR