4-Brom-3-methyl-5-(trifluormethyl)-1H-pyrazol

Lieferant: CHEMPUR

Synonyme: 4-Brom-5-methyl-3-(trifluormethyl)-pyrazol

CD07267.250 CD07267.1
CHMPCD07267.250EA 0 EUR
CHMPCD07267.250 MAYBCD07267.1
4-Brom-3-methyl-5-(trifluormethyl)-1H-pyrazol
4-Brom-5-methyl-3-(trifluormethyl)-1H-pyrazol
Formel: C₅H₄BrF₃N₂
MDL: MFCD00120266
CAS-Nummer: 60061-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR