4-Brom-m-toluylsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 4-Brom-3-methylbenzoesäure

CA-4279-25G CA-4279-100G CA-4279-5G
COBBCA-4279-25GEA 76.1 EUR
COBBCA-4279-25G COBBCA-4279-100G COBBCA-4279-5G
4-Brom-m-toluylsäure
4-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD00039526
CAS-Nummer: 7697-28-1
EINECS: 231-713-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR