4-Brom-3-methylbenzolsulfonylchlorid
Lieferant: COMBI-BLOCKS
Synonyme:
4-Brom-3-methylbenzolsulfonsäurechlorid, 4-Brom-3-methylbenzol-1-sulfonylchlorid, 4-Brom-3-methylbenzolsulfochlorid
OS-7651-1G
OS-7651-5G
OS-7651-25G
COBBOS-7651-1GEA
26.2
EUR
COBBOS-7651-1G
COBBOS-7651-5G
COBBOS-7651-25G
4-Brom-3-methylbenzolsulfonylchlorid
4-Brom-3-methylbenzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269.55 Schmelzpunkt: 61 |
MDL:
MFCD03094696 CAS-Nummer: 72256-93-0 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...