Ethyl-4-brom-3-nitrobenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-bromo-3-nitrobenzoate

CA-4539-5G CA-4539-25G CA-4539-1G
COBBCA-4539-5GEA 127 EUR
COBBCA-4539-5G COBBCA-4539-25G COBBCA-4539-1G
Ethyl-4-brom-3-nitrobenzoat
Ethyl-4-brom-3-nitrobenzoat
Formel: C₉H₈BrNO₄
Molecular Weight: 274,07 g/mol
MDL: MFCD09972120
CAS-Nummer: 168473-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR