Methyl-4-brom-3-nitrobenzoat

Lieferant: FLUOROCHEM

Synonyme: 4-Brom-3-nitrobenzoesäuremethylester

009755-1G 009755-5G
FLUO009755-1GEA 31.1 EUR
FLUO009755-1G FLUO009755-5G
Methyl-4-brom-3-nitrobenzoat
Methyl-4-brom-3-nitrobenzoat
Formel: C₈H₆BrNO₄
Molecular Weight: 260,04 g/mol
MDL: MFCD00553089
CAS-Nummer: 2363-16-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR