4-Brombenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
4-Brombenzoesäureamidin Hydrochlorid, 4-Bromphenylamidin Hydrochlorid
12-0056.1
12-0056.5
CHMP12-0056.1EA
0
EUR
CHMP12-0056.1
CHMP12-0056.5
4-Brombenzamidin Hydrochlorid
4-Brombenzamidin Hydrochlorid
Formel:
C₇H₈BrClN₂ Molecular Weight: 235.51 g/mol |
MDL:
MFCD00464967 CAS-Nummer: 55368-42-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...