4-Brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brombenzolsulfonsäureamid

OT-0094-100G OT-0094-5G OT-0094-25G
COBBOT-0094-100GEA 240 EUR
COBBOT-0094-100G COBBOT-0094-5G COBBOT-0094-25G
4-Brombenzolsulfonamid
4-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00051977
CAS-Nummer: 701-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR