4-Brombenzolsulfonamid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Brombenzolsulfonsäureamid
L08602.03
L08602.06
L08602.14
L08602.03EA
29.2
EUR
L08602.03
L08602.06
L08602.14
4-Brombenzolsulfonamid ≥98%
4-Brombenzolsulfonamid
Purity: 98%
CAS No.: 701-34-8
Molecular Formula: C6H6BrNO2S
Formula Weight: 236.09
Melting Point: 164-167°
Beilstein Reference: 2691657
Formel:
C₆H₆BrNO₂S Molecular Weight: 236.09 g/mol Schmelzpunkt: 164…167 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051977 CAS-Nummer: 701-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...