4-Bromimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Brom-1H-imidazol

HI-1024-5G HI-1024-25G HI-1024-1G
COBBHI-1024-5GEA 36.2 EUR
COBBHI-1024-5G COBBHI-1024-25G COBBHI-1024-1G
4-Bromimidazol
4-Bromoimidazole
Formel: C₃H₃BrN₂
Molecular Weight: 146,97 g/mol
MDL: MFCD00047021
CAS-Nummer: 2302-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR