4-Chlor-2,5-dimethylbenzolsulfonylchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-Chlor-2,5-dimethylbenzolsulfonsäurechlorid, 5-Chlor-p-xylol-2-sulfonsäurechlorid, 5-Chlor-p-xylol-2-sulfonylchlorid, 4-Chlor-2,5-dimethylbenzolsulfochlorid, 4-Chlor-2,5-dimethylbenzol-1-sulfonylchlorid

L05247.06 L05247.14
L05247.06EA 45.8 EUR
L05247.06 L05247.14
4-Chlor-2,5-dimethylbenzolsulfonylchlorid ≥98%
4-Chlor-2,5-dimethylbenzolsulfonylchlorid

Purity: 98% CAS No.: 88-49-3 Molecular Formula: C8H8Cl2O2S Formula Weight: 239.12 Melting Point: 48-50° Beilstein Reference: 2838165 UN No.: 3261 MDL No.: MFCD00044017

Formel: C₈H₈Cl₂O₂S
Molecular Weight: 239,12 g/mol
Schmelzpunkt: 48…50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00044017
CAS-Nummer: 88-49-3
EINECS: 201-835-5
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR