4-Chlor-2-fluorbenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

CA-4385-5G CA-4385-25G CA-4385-100G
COBBCA-4385-5GEA 25.5 EUR
COBBCA-4385-5G COBBCA-4385-25G COBBCA-4385-100G
4-Chlor-2-fluorbenzoesäure
4-Chloro-2-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00042468
CAS-Nummer: 446-30-0
EINECS: 207-162-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR