Methyl-4-chlor-2-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: 4-Chlor-2-fluorbenzoesäuremethylester

CA-4362-5G CA-4362-25G CA-4362-1G
COBBCA-4362-5GEA 86.2 EUR
COBBCA-4362-5G COBBCA-4362-25G COBBCA-4362-1G
Methyl-4-chlor-2-fluorbenzoat
Methyl-4-chlor-2-fluorbenzoat
Formel: C₈H₆ClFO₂
Molecular Weight: 188,59 g/mol
MDL: MFCD04038392
CAS-Nummer: 148893-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR