Methyl-4-chlor-2-fluorbenzoat

Lieferant: FLUOROCHEM

Synonyme: 4-Chlor-2-fluorbenzoesäuremethylester

019485-5G 019485-25G
FLUO019485-5GEA 82.1 EUR
FLUO019485-5G FLUO019485-25G
Methyl-4-chlor-2-fluorbenzoat
Methyl-4-chlor-2-fluorbenzoat
Formel: C₈H₆ClFO₂
Molecular Weight: 188.59 g/mol
MDL: MFCD04038392
CAS-Nummer: 148893-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR